2'-Fluoro[1,1'-biphenyl]-2-amine

Supplier: Matrix Scientific

021698-500MG
101812-572EA 300 USD
101812-572
2'-Fluoro[1,1'-biphenyl]-2-amine
2'-Fluoro[1,1'-biphenyl]-2-amine

Hazard: Irritant

Formula: C₁₂H₁₀FN
MDL Number: MFCD06802527
CAS Number: 316-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR