4'-Methyl[1,1'-biphenyl]-2-amine

Supplier: Matrix Scientific

021695-500MG
101812-566EA 235.75 USD
101812-566
4'-Methyl[1,1'-biphenyl]-2-amine
4'-Methyl[1,1'-biphenyl]-2-amine

Hazard: Irritant

Formula: C₁₃H₁₃N
MDL Number: MFCD03424650
CAS Number: 1204-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR