2-(2,5-Dimethoxyphenyl)piperidine

Supplier: Matrix Scientific

021690-500MG
101812-556EA 315.38 USD
101812-556
2-(2,5-Dimethoxyphenyl)piperidine
2-(2,5-Dimethoxyphenyl)piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO₂
MDL Number: MFCD02663658
CAS Number: 383128-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR