2-(2,4-Dimethoxyphenyl)piperidine

Supplier: Matrix Scientific

021689-500MG
101812-554EA 315.38 USD
101812-554
2-(2,4-Dimethoxyphenyl)piperidine
2-(2,4-Dimethoxyphenyl)piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO₂
MDL Number: MFCD02663551
CAS Number: 383128-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR