2-(4-Ethylphenyl)piperidine

Supplier: Matrix Scientific

021687-500MG
101812-550EA 235.75 USD
101812-550
2-(4-Ethylphenyl)piperidine
2-(4-Ethylphenyl)piperidine

Hazard: Irritant

Formula: C₁₃H₁₉N
MDL Number: MFCD02663673
CAS Number: 383128-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR