3-(3-Fluorophenyl)piperidine hydrochloride

Supplier: Matrix Scientific

021674-500MG
101812-524EA 186.15 USD
101812-524
3-(3-Fluorophenyl)piperidine hydrochloride
3-(3-Fluorophenyl)piperidine hydrochloride

Hazard: Irritant/Refrigerate

Formula: C₁₁H₁₅ClFN
MDL Number: MFCD02178932
CAS Number: 100240-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR