2-Phenylpyrrolidine
Supplier: Matrix Scientific
021668-500MG
101812-516EA
24.62
USD
101812-516
2-Phenylpyrrolidine
2-Phenylpyrrolidine
Hazard: Irritant
Formula:
C₁₀H₁₃N MW: 147.2200 g/mol |
MDL Number:
MFCD01631835 CAS Number: 1006-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...