2-(3,5-Dimethylphenoxy)-1-methylethylamine

Supplier: Matrix Scientific

021661-500MG
101812-502EA 370.3 USD
101812-502
2-(3,5-Dimethylphenoxy)-1-methylethylamine
2-(3,5-Dimethylphenoxy)-1-methylethylamine

Hazard: Irritant

Formula: C₁₁H₁₇NO
MDL Number: MFCD07658072
CAS Number: 117322-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR