3-(4-Ethylpiperazin-1-ylmethyl)benzoic acid

Supplier: Matrix Scientific

021656-500MG
101812-494EA 495.38 USD
101812-494
3-(4-Ethylpiperazin-1-ylmethyl)benzoic acid
3-(4-Ethylpiperazin-1-ylmethyl)benzoic acid

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₂
MDL Number: MFCD08687798
CAS Number: 919033-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR