(2-Dimethylaminomethylthiazol-4-yl)methanol hydrochloride

Supplier: Matrix Scientific

021633-500MG
101812-454EA 495.38 USD
101812-454
(2-Dimethylaminomethylthiazol-4-yl)methanol hydrochloride
(2-Dimethylaminomethylthiazol-4-yl)methanol hydrochloride

Hazard: Irritant

Formula: C₇H₁₃ClN₂OS
MDL Number: MFCD08741487
CAS Number: 82586-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR