2-(3,5-Dimethylphenoxy)ethylamine

Supplier: Matrix Scientific

021631-500MG
101812-450EA 195.5 USD
101812-450
2-(3,5-Dimethylphenoxy)ethylamine
2-(3,5-Dimethylphenoxy)ethylamine

Hazard: Irritant

Formula: C₁₀H₁₅NO
MDL Number: MFCD04061702
CAS Number: 26646-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR