(4-Methyl-2,6-dioxo-3,6-dihydro-2H-pyrimidin-1-yl)acetic acid

Supplier: Matrix Scientific

021629-500MG
101812-446EA 186.15 USD
101812-446
(4-Methyl-2,6-dioxo-3,6-dihydro-2H-pyrimidin-1-yl)acetic acid
(4-Methyl-2,6-dioxo-3,6-dihydro-2H-pyrimidin-1-yl)acetic acid

Hazard: Irritant

Formula: C₇H₈N₂O₄
MDL Number: MFCD01326529
CAS Number: 54069-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR