1-(3-Aminopropyl)piperidine-4-carboxylic acidamide

Supplier: Matrix Scientific

021628-500MG
101812-444EA 495.38 USD
101812-444
1-(3-Aminopropyl)piperidine-4-carboxylic acidamide
1-(3-Aminopropyl)piperidine-4-carboxylic acidamide

Hazard: Irritant

Formula: C₉H₁₉N₃O
MDL Number: MFCD08686836
CAS Number: 4608-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR