2-(4-Methoxyphenyl)pyrrolidine

Supplier: Matrix Scientific

Synonyms: 4-(2-Pyrrolidinyl)anisole

021627-500MG
101812-442EA 185.15 USD
101812-442
2-(4-Methoxyphenyl)pyrrolidine
2-(4-Methoxyphenyl)pyrrolidine

Hazard: Irritant

Formula: C₁₁H₁₅NO
MDL Number: MFCD01631836
CAS Number: 74190-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR