5-(2-Chloroacetyl)-1,3-dihydrobenzoimidazol-2-one

021620-500MG
101812-434EA 321 USD
101812-434
5-(2-Chloroacetyl)-1,3-dihydrobenzoimidazol-2-one
5-(2-Chloroacetyl)-1,3-dihydrobenzoimidazol-2-one

Hazard: Irritant

Formula: C₉H₇ClN₂O₂
MW: 210.62 g/mol
MDL Number: MFCD04624846
CAS Number: 93202-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR