1-(2-Ethoxyethyl)piperazine

Supplier: Matrix Scientific

021601-500MG
101812-400EA 280 USD
101812-400
1-(2-Ethoxyethyl)piperazine
1-(2-Ethoxyethyl)piperazine

Hazard: Irritant

Formula: C₈H₁₈N₂O
MDL Number: MFCD01320899
CAS Number: 13484-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR