2-Dimethylamino-4-methoxythiazole-5-carbaldehyde

Supplier: Matrix Scientific

021598-500MG
101812-392EA 261.54 USD
101812-392
2-Dimethylamino-4-methoxythiazole-5-carbaldehyde
2-Dimethylamino-4-methoxythiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₇H₁₀N₂O₂S
Melting Pt: 112 °C
MDL Number: MFCD07852054
CAS Number: 877977-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR