Methyl-2-amino-4-methylthiazole-5-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-4-methylthiazole-5-carboxylate

021583-500MG
101812-362EA 13.8 USD
101812-362
Methyl-2-amino-4-methylthiazole-5-carboxylate
Methyl 2-amino-4-methylthiazole-5-carboxylate

Hazard: Irritant

Formula: C₆H₈N₂O₂S
MW: 172.2049 g/mol
MDL Number: MFCD01993690
CAS Number: 3829-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR