2-Methoxy-3-methylbenzoic acid

Supplier: Matrix Scientific

021575-500MG
101812-344EA 376.92 USD
101812-344
2-Methoxy-3-methylbenzoic acid
2-Methoxy-3-methylbenzoic acid

Hazard: Irritant

Formula: C₉H₁₀O₃
MDL Number: MFCD00016522
CAS Number: 26507-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR