(1-Aminomethyl-4-methylcyclohexyl)dimethylamine
Supplier: Matrix Scientific
021569-500MG
101812-334EA
253.85
USD
101812-334
(1-Aminomethyl-4-methylcyclohexyl)dimethylamine
(1-Aminomethyl-4-methylcyclohexyl)dimethylamine
Hazard: Irritant
Formula:
C₁₀H₂₂N₂ |
MDL Number:
MFCD08667802 CAS Number: 919013-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...