5-Cyclopropyl-1-(1,1-dioxotetrahydro-1λ*6*-thiophen-3-yl)-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

021562-500MG
101812-320EA 261.54 USD
101812-320
5-Cyclopropyl-1-(1,1-dioxotetrahydro-1λ*6*-thiophen-3-yl)-1H-pyrazole-3-carboxylic acid
5-Cyclopropyl-1-(1,1-dioxotetrahydro-1lambda*6*-thiophen-3-yl)-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₄S
MW: 270.31 g/mol
MDL Number: MFCD08568559
CAS Number: 956687-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR