1-Isobutyl-3-methyl-1H-pyrazole-4-sulfonyl chloride

Supplier: Matrix Scientific

021555-500MG
101812-306EA 269.23 USD
101812-306
1-Isobutyl-3-methyl-1H-pyrazole-4-sulfonyl chloride
1-Isobutyl-3-methyl-1H-pyrazole-4-sulfonyl chloride

Hazard: Irritant

Formula: C₈H₁₃ClN₂O₂S
MDL Number: MFCD06805445
CAS Number: 1006453-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR