1-Isopropyl-3-methyl-1H-pyrazole-4-sulfonyl chloride

Supplier: Matrix Scientific

021554-500MG
101812-304EA 261.54 USD
101812-304
1-Isopropyl-3-methyl-1H-pyrazole-4-sulfonyl chloride
1-Isopropyl-3-methyl-1H-pyrazole-4-sulfonyl chloride

Hazard: Irritant

Formula: C₇H₁₁ClN₂O₂S
MDL Number: MFCD06805443
CAS Number: 1006453-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR