2-(4-Aminophenoxy)-1-pyrrolidin-1-ylethanone

Supplier: Matrix Scientific

021550-500MG
101812-296EA 495.38 USD
101812-296
2-(4-Aminophenoxy)-1-pyrrolidin-1-ylethanone
2-(4-Aminophenoxy)-1-pyrrolidin-1-ylethanone

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD06662315
CAS Number: 76870-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR