2-(4-Methoxyphenoxy)-2-methylpropionic acid

Supplier: Matrix Scientific

021538-500MG
101812-272EA 280 USD
101812-272
2-(4-Methoxyphenoxy)-2-methylpropionic acid
2-(4-Methoxyphenoxy)-2-methylpropionic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₄
MDL Number: MFCD03422284
CAS Number: 17509-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR