1-Methyl-2-piperidin-1-ylethylamine

Supplier: Matrix Scientific

021536-500MG
101812-268EA 370.3 USD
101812-268
1-Methyl-2-piperidin-1-ylethylamine
1-Methyl-2-piperidin-1-ylethylamine

Hazard: Irritant

Formula: C₈H₁₈N₂
Boiling Pt: 199.5 °C (760 mmHg)
Density: 0.913
Flash Pt: 70.9 °C
MDL Number: MFCD01318922
CAS Number: 34217-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR