2-(4-Fluorophenoxy)-2-methylpropionic acid

Supplier: Matrix Scientific

021535-500MG
101812-266EA 240.35 USD
101812-266
2-(4-Fluorophenoxy)-2-methylpropionic acid
2-(4-Fluorophenoxy)-2-methylpropionic acid

Hazard: Irritant

Formula: C₁₀H₁₁FO₃
MDL Number: MFCD00093187
CAS Number: 587-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR