2-Aminomethyladamantan-2-ol

Supplier: Matrix Scientific

021525-500MG
101812-248EA 201.25 USD
101812-248
2-Aminomethyladamantan-2-ol
2-Aminomethyladamantan-2-ol

Hazard: Irritant

Formula: C₁₁H₁₉NO
MDL Number: MFCD01837514
CAS Number: 24779-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR