Azepan-1-ylpiperidin-3-ylmethanonetrifluoroacetate

Supplier: Matrix Scientific

021522-500MG
101812-242EA 186.15 USD
101812-242
Azepan-1-ylpiperidin-3-ylmethanonetrifluoroacetate
Azepan-1-ylpiperidin-3-ylmethanonetrifluoroacetate

Hazard: Irritant

Formula: C₁₄H₂₃F₃N₂O₃
MW: 324.35 g/mol
MDL Number: MFCD08741478
CAS Number: 1185052-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR