C-(4-m-Tolyltetrahydropyran-4-yl)methylamine

Supplier: Matrix Scientific

021518-500MG
101812-234EA 495.38 USD
101812-234
C-(4-m-Tolyltetrahydropyran-4-yl)methylamine
C-(4-m-Tolyltetrahydropyran-4-yl)methylamine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD08688551
CAS Number: 917747-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR