2-Pyrrolidin-1-ylpropylamine

Supplier: Matrix Scientific

021517-500MG
101812-232EA 269.23 USD
101812-232
2-Pyrrolidin-1-ylpropylamine
2-Pyrrolidin-1-ylpropylamine

Hazard: Irritant

Formula: C₇H₁₆N₂
MDL Number: MFCD06446933
CAS Number: 50998-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR