2-(4-Methylpiperazin-1-yl)propylamine

Supplier: Matrix Scientific

021516-500MG
101812-230EA 269.23 USD
101812-230
2-(4-Methylpiperazin-1-yl)propylamine
2-(4-Methylpiperazin-1-yl)propylamine

Hazard: Irritant

Formula: C₈H₁₉N₃
MDL Number: MFCD06446939
CAS Number: 70717-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR