2-(2-Methyl-piperidine-1-sulfonyl)-ethylamineoxalate

Supplier: Matrix Scientific

021512-500MG
101812-222EA 195.5 USD
101812-222
2-(2-Methyl-piperidine-1-sulfonyl)-ethylamineoxalate
2-(2-Methyl-piperidine-1-sulfonyl)-ethylamineoxalate

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₆S
MDL Number: MFCD08741495
CAS Number: 1185304-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR