C-(4-Fluorophenyl)-C-(1-methyl-1H-imidazol-2-yl)methylamine

Supplier: Matrix Scientific

021509-500MG
101812-216EA 307.69 USD
101812-216
C-(4-Fluorophenyl)-C-(1-methyl-1H-imidazol-2-yl)methylamine
C-(4-Fluorophenyl)-C-(1-methyl-1H-imidazol-2-yl)methylamine

Hazard: Irritant

Formula: C₁₁H₁₂FN₃
MDL Number: MFCD07686047
CAS Number: 872107-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR