3-Imidazol-1-ylmethylpiperidine hydrochloride

Supplier: Matrix Scientific

021508-500MG
101812-214EA 269.23 USD
101812-214
3-Imidazol-1-ylmethylpiperidine hydrochloride
3-Imidazol-1-ylmethylpiperidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₆ClN₃
MDL Number: MFCD08741490
CAS Number: 1185293-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR