2,2-Dimethylchroman-4-ylamine

Supplier: Matrix Scientific

021506-500MG
101812-210EA 201.25 USD
101812-210
2,2-Dimethylchroman-4-ylamine
2,2-Dimethylchroman-4-ylamine

Hazard: Irritant

Formula: C₁₁H₁₅NO
MDL Number: MFCD08687615
CAS Number: 220634-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR