2-Acetylaminothiazole-5-sulfonyl chloride

Supplier: Matrix Scientific

021505-500MG
101812-208EA 415.38 USD
101812-208
2-Acetylaminothiazole-5-sulfonyl chloride
2-Acetylaminothiazole-5-sulfonyl chloride

Hazard: Irritant

Formula: C₅H₅ClN₂O₃S₂
Melting Pt: 210 °C (dec.)
MDL Number: MFCD05022486
CAS Number: 69812-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR