4-Methoxy-2,5-dimethyl-benzenesulfonyl chloride

Supplier: Matrix Scientific

021499-500MG
101812-198EA 269.23 USD
101812-198
4-Methoxy-2,5-dimethyl-benzenesulfonyl chloride
4-Methoxy-2,5-dimethyl-benzenesulfonyl chloride

Hazard: Irritant

Formula: C₉H₁₁ClO₃S
MDL Number: MFCD08688025
CAS Number: 91179-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR