C-[1-(2-Methoxyethyl)pyrrolidin-3-yl]methylamine

Supplier: Matrix Scientific

021488-500MG
101812-178EA 158.46 USD
101812-178
C-[1-(2-Methoxyethyl)pyrrolidin-3-yl]methylamine
C-[1-(2-Methoxyethyl)pyrrolidin-3-yl]methylamine

Hazard: Irritant

Formula: C₈H₁₈N₂O
MDL Number: MFCD08059731
CAS Number: 910443-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR