2-[(4-Chlorophenyl)sulfonyl]acetic acid

Supplier: Matrix Scientific

021485-500MG
101812-172EA 161.54 USD
101812-172
2-[(4-Chlorophenyl)sulfonyl]acetic acid
2-[(4-Chlorophenyl)sulfonyl]acetic acid

Hazard: Irritant

Formula: C₈H₇ClO₄S
MDL Number: MFCD00201948
CAS Number: 3405-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR