2,2-Dimethylchroman-6-sulfonyl chloride
Supplier: Matrix Scientific
021484-500MG
101812-170EA
256.92
USD
101812-170
2,2-Dimethylchroman-6-sulfonyl chloride
2,2-Dimethylchroman-6-sulfonyl chloride
Hazard: Irritant
Formula:
C₁₁H₁₃ClO₃S |
MDL Number:
MFCD03407324 CAS Number: 131880-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...