1-Dimethylsulfamoylpyrrolidine-2-carboxylic acid
Supplier: Matrix Scientific
021445-500MG
101812-126EA
495.38
USD
101812-126
1-Dimethylsulfamoylpyrrolidine-2-carboxylic acid
1-Dimethylsulfamoylpyrrolidine-2-carboxylic acid
Hazard: Irritant
Formula:
C₇H₁₄N₂O₄S |
MDL Number:
MFCD01025468 CAS Number: 1104518-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...