(4-Isopropyl-[1,2,3]-triazol-1-yl)acetic acid

Supplier: Matrix Scientific

021358-500MG
101811-842EA 370.3 USD
101811-842
(4-Isopropyl-[1,2,3]-triazol-1-yl)acetic acid
(4-Isopropyl-[1,2,3]-triazol-1-yl)acetic acid

Hazard: Irritant

Formula: C₇H₁₁N₃O₂
MDL Number: MFCD08445772
CAS Number: 887405-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR