1-(2-Isopropoxyethyl)piperazine

Supplier: Matrix Scientific

021357-500MG
101811-840EA 164.62 USD
101811-840
1-(2-Isopropoxyethyl)piperazine
1-(2-Isopropoxyethyl)piperazine

Hazard: Irritant

Formula: C₉H₂₀N₂O
MDL Number: MFCD08445769
CAS Number: 889939-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR