2-Azepan-1-yl-2-furan-2-ylethylamine

Supplier: Matrix Scientific

021350-500MG
101811-826EA 195.5 USD
101811-826
2-Azepan-1-yl-2-furan-2-ylethylamine
2-Azepan-1-yl-2-furan-2-ylethylamine

Hazard: Irritant

Formula: C₁₂H₂₀N₂O
MDL Number: MFCD08361863
CAS Number: 889939-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR