2-(4-Chlorophenyl)-2-morpholin-4-ylethylamine

Supplier: Matrix Scientific

021334-500MG
101811-794EA 209.3 USD
101811-794
2-(4-Chlorophenyl)-2-morpholin-4-ylethylamine
2-(4-Chlorophenyl)-2-morpholin-4-ylethylamine

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD08437446
CAS Number: 866782-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR