(1-Methylpiperidin-4-yl)(2-morpholin-4-ylethyl)amine

Supplier: Matrix Scientific

021332-500MG
101811-790EA 370.3 USD
101811-790
(1-Methylpiperidin-4-yl)(2-morpholin-4-ylethyl)amine
(1-Methylpiperidin-4-yl)(2-morpholin-4-ylethyl)amine

Hazard: Irritant

Formula: C₁₂H₂₅N₃O
MDL Number: MFCD00979774
CAS Number: 416865-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR