2-(3,5-Dimethyladamantan-1-yloxy)ethylamine

Supplier: Matrix Scientific

021316-500MG
101811-762EA 370.3 USD
101811-762
2-(3,5-Dimethyladamantan-1-yloxy)ethylamine
2-(3,5-Dimethyladamantan-1-yloxy)ethylamine

Hazard: Irritant

Formula: C₁₄H₂₅NO
MDL Number: MFCD08445731
CAS Number: 887405-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR