C-(5-Methyl[1,2,4]oxadiazol-3-yl)methylamine hydrochloride

Supplier: Matrix Scientific

Synonyms: (5-Methyl-1,2,4-oxadiazol-3-yl)methanamine hydrochloride

021314-500MG
101811-758EA 209.3 USD
101811-758
C-(5-Methyl[1,2,4]oxadiazol-3-yl)methylamine hydrochloride
C-(5-Methyl[1,2,4]oxadiazol-3-yl)methylamine hydrochloride

Hazard: Irritant

Formula: C₄H₈ClN₃O
MW: 149.58 g/mol
MDL Number: MFCD11506367
CAS Number: 1184986-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR