C-(4-p-Tolyltetrahydropyran-4-yl)methylamine

Supplier: Matrix Scientific

021312-500MG
101811-754EA 370.3 USD
101811-754
C-(4-p-Tolyltetrahydropyran-4-yl)methylamine
C-(4-p-Tolyltetrahydropyran-4-yl)methylamine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD08445817
CAS Number: 619296-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR